This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05962189_m1
See other LOC100394788 GE Assays ›
Gene Symbol:
LOC100394788
Gene Name:
histone-lysine N-methyltransferase SETDB2
Gene Aliases:
SETDB2
Chromosome Location:
Chr.1: 65338055 - 65395212 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05962189_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100394788
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_008991860.2 XP_008990108.1 6 - 7 857 100
XM_008991864.2 XP_008990112.1 6 - 7 857 100
XM_008991868.2 XP_008990116.1 6 - 7 878 100
XM_008991872.2 XP_008990120.1 4 - 5 783 100
XM_008991877.2 XP_008990125.1 5 - 6 1335 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0