Assay Details
Gene Symbol: LOC100394788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008991860.2 | XP_008990108.1 | 4 - 5 | 700 | 93 | |
XM_008991864.2 | XP_008990112.1 | 4 - 5 | 700 | 93 | ||
XM_008991868.2 | XP_008990116.1 | 4 - 5 | 721 | 93 | ||
XM_008991872.2 | XP_008990120.1 | 2 - 3 | 626 | 93 | ||
XM_008991877.2 | XP_008990125.1 | 3 - 4 | 1178 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
