Assay Details
Gene Symbol: ARL6IP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749434.3 | XP_002749480.1 | 1 - 2 | 975 | 76 | |
XM_008998887.2 | XP_008997135.1 | 1 - 2 | 975 | 76 | ||
XM_008998889.2 | XP_008997137.1 | 1 - 2 | 975 | 76 | ||
XM_017973478.1 | XP_017828967.1 | 1 - 2 | 975 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
