This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05951499_m1
See other ITPA GE Assays ›
Gene Symbol:
ITPA
Gene Name:
inosine triphosphatase
Gene Aliases:
-
Chromosome Location:
Chr.5: 9006011 - 9021207 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05951499_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ITPA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002747405.4 XP_002747451.3 7 - 8 572 77
XM_017971917.1 XP_017827406.1 8 - 9 640 77
XM_017971918.1 XP_017827407.1 6 - 7 509 77
XM_017971919.1 XP_017827408.1 6 - 7 536 77
XM_017971920.1 XP_017827409.1 6 - 7 464 77
XM_017971922.1 XP_017827411.1 5 - 6 477 77
XM_017971923.1 XP_017827412.1 7 - 8 689 77
XM_017971924.1 XP_017827413.1 7 - 8 494 77
XM_017971925.1 XP_017827414.1 7 - 8 497 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

nucleotide phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0