This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05950061_g1
See other TRIM56 GE Assays ›
Gene Symbol:
TRIM56
Gene Name:
tripartite motif containing 56
Gene Aliases:
-
Chromosome Location:
Chr.2: 10955284 - 10960341 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05950061_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRIM56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002743972.4 XP_002744018.1 2 - 3 513 85
XM_008982178.2 XP_008980426.1 2 - 3 254 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_002743972.4, XM_008982178.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0