This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05942956_m1
See other GHRL GE Assays ›
Gene Symbol:
GHRL
Gene Name:
ghrelin and obestatin prepropeptide
Gene Aliases:
MTLRP
Chromosome Location:
Chr.15: 62210548 - 62218403 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05942956_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GHRL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017965077.1 XP_017820566.1 2 - 3 730 86
XM_017965078.1 XP_017820567.1 2 - 3 710 86
XM_017965079.1 XP_017820568.1 3 - 4 3357 86
XM_017965080.1 XP_017820569.1 3 - 4 635 86
XM_017965081.1 XP_017820570.1 3 - 4 3344 86
GenBank mRNA LC145038.1 - 2 - 3 227 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

peptide hormone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0