This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05925671_m1
See other HDAC11 GE Assays ›
Gene Symbol:
HDAC11
Gene Name:
histone deacetylase 11
Gene Aliases:
-
Chromosome Location:
Chr.15: 65421392 - 65448084 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05925671_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002758648.4 XP_002758694.2 7 - 8 666 70
XM_008982179.2 XP_008980427.1 7 - 8 1087 70
XM_008982181.2 XP_008980429.1 7 - 8 607 70
XM_017965110.1 XP_017820599.1 4 - 5 850 70
XM_017965111.1 XP_017820600.1 7 - 8 1087 70
XM_017965112.1 XP_017820601.1 5 - 6 909 70
XM_017965113.1 XP_017820602.1 7 - 8 638 70
XM_017965114.1 XP_017820603.1 7 - 8 601 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0