Assay Details
Gene Symbol: PDPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008986200.2 | XP_008984448.1 | 18 | 4567 | 95 | |
XM_008986201.2 | XP_008984449.1 | 17 | 4477 | 95 | ||
XM_017966965.1 | XP_017822454.1 | 16 | 4526 | 95 | ||
XM_017966966.1 | XP_017822455.1 | 15 | 4003 | 95 | ||
XM_017966967.1 | XP_017822456.1 | 10 | 3243 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
