This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj05919388_m1
See other CLDND1 GE Assays ›
Gene Symbol:
CLDND1
Gene Name:
claudin domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.15: 98383131 - 98391151 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj05919388_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CLDND1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002758873.4 XP_002758919.3 3 - 4 575 115
XM_008982586.2 XP_008980834.1 2 - 3 547 115
XM_008982587.2 XP_008980835.1 3 - 4 573 115
XM_008982588.2 XP_008980836.1 3 - 4 519 115
XM_008982589.2 XP_008980837.1 3 - 4 666 115
XM_008982590.2 XP_008980838.1 3 - 4 423 115
XM_008982591.2 XP_008980839.1 3 - 4 436 115
XM_008982592.2 XP_008980840.1 3 - 4 390 115
XM_008982593.2 XP_008980841.1 3 - 4 412 115
XM_017965296.1 XP_017820785.1 3 - 4 575 115
XM_017965297.1 XP_017820786.1 3 - 4 430 115
XM_017965298.1 XP_017820787.1 3 - 4 353 115
XM_017965299.1 XP_017820788.1 3 - 4 325 115
XM_017965300.1 XP_017820789.1 2 - 3 387 115

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0