This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj01568268_m1
See other APC GE Assays ›
Gene Symbol:
APC
Gene Name:
APC, WNT signaling pathway regulator
Gene Aliases:
-
Chromosome Location:
Chr.2: 94111037 - 94262957 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj01568268_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002744711.3 XP_002744757.1 13 - 14 1709 72
XM_008991687.2 XP_008989935.1 13 - 14 1935 72
XM_008991688.2 XP_008989936.1 14 - 15 2086 72
XM_008991689.2 XP_008989937.1 14 - 15 1763 72
XM_008991691.2 XP_008989939.1 12 - 13 1880 72
XM_008991692.2 XP_008989940.1 13 - 14 2031 72
XM_008991696.2 XP_008989944.1 13 - 14 1629 72
XM_008991697.2 XP_008989945.1 13 - 14 1724 72
XM_017969743.1 XP_017825232.1 13 - 14 2490 72
XM_017969744.1 XP_017825233.1 15 - 16 1824 72
XM_017969745.1 XP_017825234.1 14 - 15 1812 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0