Assay Details
Gene Symbol: RCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017974365.1 | XP_017829854.1 | 6 - 7 | 922 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |

Gene Symbol: RCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017974365.1 | XP_017829854.1 | 6 - 7 | 922 | 83 |
Set Membership: |
![]() ![]() ![]() |