Assay Details
Gene Symbol: CTNNA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017964211.1 | XP_017819700.1 | 10 - 11 | 1599 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
