Assay Details
Gene Symbol: SAMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017967456.1 | XP_017822945.1 | 10 - 11 | 2708 | 78 | |
XM_017967457.1 | XP_017822946.1 | 11 - 12 | 2388 | 78 | ||
XM_017967458.1 | XP_017822947.1 | 11 - 12 | 2389 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
