Assay Details
Gene Symbol: GPR137B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760885.3 | XP_002760931.1 | 1 - 2 | 529 | 87 | |
XM_017966722.1 | XP_017822211.1 | 1 - 2 | 529 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
