Assay Details
Gene Symbol: MFSD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008999092.2 | XP_008997340.1 | 3 - 4 | 2919 | 57 | |
XM_008999093.2 | XP_008997341.1 | 4 - 5 | 2041 | 57 | ||
XM_008999094.2 | XP_008997342.1 | 4 - 5 | 1924 | 57 | ||
XM_008999095.2 | XP_008997343.1 | 4 - 5 | 1882 | 57 | ||
XM_008999096.2 | XP_008997344.1 | 3 - 4 | 2919 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
