Assay Details
Gene Symbol: ECM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008984270.2 | XP_008982518.1 | 8 - 9 | 1906 | 72 | |
XM_008984271.2 | XP_008982519.1 | 7 - 8 | 1804 | 72 | ||
XM_017966243.1 | XP_017821732.1 | 7 - 8 | 1531 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
