This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cj00924121_g1
See other PCBP4 GE Assays ›
Gene Symbol:
PCBP4
Gene Name:
poly(rC) binding protein 4
Gene Aliases:
-
Chromosome Location:
Chr.15: 27703559 - 27714541 on Build Callithrix_jacchus_cj1700_1.1
Species:
Callithrix jacchus
Species Specific ID (Flybase ID):
-
Assay ID Cj00924121_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCBP4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_002758401.4 XP_002758447.1 4 - 5 481 56
XM_002758403.4 XP_002758449.1 3 - 4 333 56
XM_008981761.2 XP_008980009.1 4 - 5 481 56
XM_008981762.2 XP_008980010.1 3 - 4 333 56

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_008981761.2, XM_008981762.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0