Assay Details
Gene Symbol: GAREM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757156.3 | XP_002757202.1 | 3 - 4 | 537 | 95 | |
XM_008979663.2 | XP_008977911.1 | 3 - 4 | 539 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
