This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04807998_gH
See other IGBP1 GE Assays ›
Gene Symbol:
IGBP1
Gene Name:
immunoglobulin binding protein 1
Gene Aliases:
-
Chromosome Location:
Chr.X|NW_017189516: 11436411 - 11481693 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04807998_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGBP1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005700707.3 XP_005700764.3 1 - 2 711 88
XM_018043890.1 XP_017899379.1 1 - 2 711 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header