This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04797730_m1
See other LOC102172692 GE Assays ›
Gene Symbol:
LOC102172692
Gene Name:
phosphatidylinositol phosphatase PTPRQ
Gene Aliases:
-
Chromosome Location:
Chr.5: 9791446 - 10035602 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04797730_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102172692
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018047601.1 XP_017903090.1 38 - 39 86
GenBank mRNA JO426300.1 - 7 - 8 1015 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 G-protein coupled receptor Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0