This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04796876_m1
See other SDR42E1 GE Assays ›
Gene Symbol:
SDR42E1
Gene Name:
short chain dehydrogenase/reductase family 42E, member 1
Gene Aliases:
-
Chromosome Location:
Chr.18: 9883975 - 9897660 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04796876_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDR42E1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005691827.3 XP_005691884.2 2 - 3 193 70
XM_005691829.3 XP_005691886.2 2 - 3 188 70
XM_013970957.2 XP_013826411.2 3 - 4 1514 70
XM_013970960.2 XP_013826414.2 2 - 3 261 70
XM_018061767.1 XP_017917256.1 3 - 4 1280 70
XM_018061768.1 XP_017917257.1 3 - 4 317 70
GenBank mRNA JO428258.1 - 2 - 3 247 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

dehydratase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header