This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04792403_m1
See other LOC102173760 GE Assays ›
Gene Symbol:
LOC102173760
Gene Name:
fer-1-like protein 4
Gene Aliases:
FER1L4
Chromosome Location:
Chr.13: 64365148 - 64404127 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04792403_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102173760
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018057769.1 XP_017913258.1 32 - 33 4060 77
XM_018057770.1 XP_017913259.1 32 - 33 4060 77
XM_018057771.1 XP_017913260.1 31 - 32 3907 77
XM_018057772.1 XP_017913261.1 32 - 33 4060 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header