This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04787222_m1
See other WASF2 GE Assays ›
Gene Symbol:
WASF2
Gene Name:
WASP family member 2
Gene Aliases:
-
Chromosome Location:
Chr.2: 10083758 - 10157651 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04787222_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: WASF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013973964.2 XP_013829418.2 8 - 9 1411 65
XM_018056745.1 XP_017912234.1 8 - 9 1554 65
XM_018056751.1 XP_017912240.1 8 - 9 1471 65
XM_018056759.1 XP_017912248.1 8 - 9 1625 65
GenBank mRNA KA336141.1 - 1 - 2 172 65

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

non-motor actin binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0