This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04785272_m1
See other PPWD1 GE Assays ›
Gene Symbol:
PPWD1
Gene Name:
peptidylprolyl isomerase domain and WD repeat containing 1
Gene Aliases:
-
Chromosome Location:
Chr.20: 13974167 - 13995328 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04785272_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPWD1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005694635.3 XP_005694692.1 8 - 9 1593 69
XM_013972735.2 XP_013828189.1 7 - 8 1490 69
GenBank mRNA JO397039.2 - 4 - 5 388 69
JO527171.3 - 4 - 5 390 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

chaperone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0