Assay Details
Gene Symbol: CREB3L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677475.3 | XP_005677532.2 | 6 - 7 | 2123 | 61 | |
XM_005677477.3 | XP_005677534.2 | 7 - 8 | 1980 | 61 | ||
XM_005677478.3 | XP_005677535.2 | 7 - 8 | 1977 | 61 | ||
XM_005677479.3 | XP_005677536.2 | 7 - 8 | 1977 | 61 | ||
XM_005677480.3 | XP_005677537.2 | 9 - 10 | 1661 | 61 | ||
XM_005677481.3 | XP_005677538.2 | 9 - 10 | 1658 | 61 | ||
XM_013977116.2 | XP_013832570.2 | 7 - 8 | 2005 | 61 | ||
XM_013977123.2 | XP_013832577.2 | 8 - 9 | 1808 | 61 | ||
XM_018046152.1 | XP_017901641.1 | 7 - 8 | 1925 | 61 | ||
XM_018046153.1 | XP_017901642.1 | 6 - 7 | 2120 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
