This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04782199_m1
See other BACH1 GE Assays ›
Gene Symbol:
BACH1
Gene Name:
BTB domain and CNC homolog 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 5168613 - 5214987 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04782199_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BACH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018049134.1 XP_017904623.1 3 - 4 1708 74
XM_018049175.1 XP_017904664.1 3 - 4 1779 74
GenBank mRNA JO330223.2 - 2 - 3 923 74
JO330224.2 - 2 - 3 923 74
JO330225.2 - 2 - 3 923 74
JO368717.2 - 2 - 3 1507 74
JO382966.2 - 2 - 3 574 74
JO427316.1 - 2 - 3 970 74
JO427541.1 - 2 - 3 970 74
JO427617.1 - 2 - 3 970 74
JO437719.2 - 2 - 3 910 74
JO499335.2 - 1 - 2 765 74
JO502051.2 - 1 - 2 765 74
JO540118.2 - 1 - 2 1083 74
JO540119.2 - 1 - 2 1083 74
JO595488.2 - 2 - 3 903 74
JP351375.2 - 1 - 2 1106 74
JP351376.2 - 1 - 2 1106 74
JP351377.2 - 1 - 2 1106 74
JT658611.1 - 2 - 3 401 74
JT705928.1 - 2 - 3 709 74
JT737981.1 - 2 - 3 702 74
KA341110.1 - 3 - 4 1753 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0