This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04781140_m1
See other AURKB GE Assays ›
Gene Symbol:
AURKB
Gene Name:
aurora kinase B
Gene Aliases:
-
Chromosome Location:
Chr.19: 27571804 - 27576960 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04781140_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AURKB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005693548.3 XP_005693605.1 7 - 8 820 88
XM_005693549.3 XP_005693606.1 6 - 7 748 88
XM_013972593.2 XP_013828047.1 7 - 8 808 88
XM_018064617.1 XP_017920106.1 7 - 8 830 88
XM_018064618.1 XP_017920107.1 6 - 7 758 88
XM_018064619.1 XP_017920108.1 4 - 5 556 88
GenBank mRNA JO598601.2 - 2 - 3 1097 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0