Assay Details
Gene Symbol: LOC102184306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040393.1 | XP_017895882.1 | 8 - 9 | 740 | 63 | |
XM_018040394.1 | XP_017895883.1 | 8 - 9 | 740 | 63 | ||
GenBank mRNA | JO635855.2 | - | 8 - 9 | 895 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
