This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04775644_m1
See other LOC102184306 GE Assays ›
Gene Symbol:
LOC102184306
Gene Name:
ras GTPase-activating protein 4
Gene Aliases:
RASA4
Chromosome Location:
Chr.25: 35461638 - 35484302 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04775644_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102184306
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018040393.1 XP_017895882.1 9 - 10 852 68
XM_018040394.1 XP_017895883.1 9 - 10 852 68
GenBank mRNA JO355430.2 - 1 - 2 81 68
JO635855.2 - 9 - 10 1007 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0