Assay Details
Gene Symbol: RD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059965.1 | XP_017915454.1 | 1 - 2 | 293 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: RD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059965.1 | XP_017915454.1 | 1 - 2 | 293 | 103 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |