This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04772327_m1
See other S100PBP GE Assays ›
Gene Symbol:
S100PBP
Gene Name:
S100P binding protein
Gene Aliases:
-
Chromosome Location:
Chr.2: 14870993 - 14904463 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04772327_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100PBP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013973507.2 XP_013828961.2 4 - 5 3423 75
XM_013973511.2 XP_013828965.2 3 - 4 2176 75
XM_018057928.1 XP_017913417.1 5 - 6 1209 75
XM_018057934.1 XP_017913423.1 4 - 5 1209 75
XM_018057941.1 XP_017913430.1 4 - 5 1177 75
XM_018057956.1 XP_017913445.1 5 - 6 1221 75
XM_018057963.1 XP_017913452.1 4 - 5 1210 75
XM_018057979.1 XP_017913468.1 5 - 6 1178 75
XM_018057990.1 XP_017913479.1 4 - 5 1180 75
XM_018057998.1 XP_017913487.1 3 - 4 1031 75
XM_018058005.1 XP_017913494.1 3 - 4 1034 75
XM_018058010.1 XP_017913499.1 5 - 6 1176 75
XM_018058016.1 XP_017913505.1 3 - 4 1406 75
XM_018058025.1 XP_017913514.1 3 - 4 290 75
GenBank mRNA JO585251.2 - 3 - 4 896 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0