Assay Details
Gene Symbol: EEF2KMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040850.1 | XP_017896339.1 | 7 - 8 | 1007 | 70 | |
XM_018040851.1 | XP_017896340.1 | 5 - 6 | 824 | 70 | ||
XM_018040852.1 | XP_017896341.1 | 4 - 5 | 479 | 70 | ||
GenBank mRNA | JO396475.2 | - | 1 - 2 | 794 | 70 | |
JO603679.2 | - | 1 - 2 | 767 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
