This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04770216_m1
See other TMBIM7P GE Assays ›
Gene Symbol:
TMBIM7P
Gene Name:
transmembrane BAX inhibitor motif containing 7, pseudogene
Gene Aliases:
-
Chromosome Location:
Chr.4: 110850279 - 110869325 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04770216_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMBIM7P
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_013963124.1 XP_013818578.1 2 - 3 351 86
XM_018047434.1 XP_017902923.1 2 - 3 349 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0