Assay Details
Gene Symbol: HNRNPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685369.3 | XP_005685426.1 | 4 - 5 | 570 | 87 | |
XM_013966923.2 | XP_013822377.1 | 4 - 5 | 570 | 87 | ||
XM_013966924.2 | XP_013822378.1 | 5 - 6 | 615 | 87 | ||
XM_013966925.2 | XP_013822379.1 | 3 - 4 | 544 | 87 | ||
XM_013966926.2 | XP_013822380.1 | 3 - 4 | 1001 | 87 | ||
XM_013966928.2 | XP_013822382.1 | 3 - 4 | 1008 | 87 | ||
XM_018054547.1 | XP_017910036.1 | 5 - 6 | 734 | 87 | ||
XM_018054548.1 | XP_017910037.1 | 6 - 7 | 779 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
