This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04768379_m1
See other PKM GE Assays ›
Gene Symbol:
PKM
Gene Name:
pyruvate kinase M1/2
Gene Aliases:
PKM2
Chromosome Location:
Chr.10: 82238789 - 82265544 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04768379_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PKM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005685176.3 XP_005685233.1 3 - 4 487 69
XM_005685177.3 XP_005685234.1 3 - 4 477 69
GenBank mRNA JR119393.1 - 5823 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0