This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04760047_m1
See other LOC102183007 GE Assays ›
Gene Symbol:
LOC102183007
Gene Name:
cell surface glycoprotein CD200 receptor 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 57399891 - 57440293 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04760047_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102183007
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018066089.1 XP_017921578.1 3 - 4 441 75
GenBank mRNA JO320926.2 - 3 - 4 635 75
JO545241.2 - 3 - 4 403 75
JO603598.2 - 3 - 4 429 75
KA306746.1 - 3 - 4 634 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0