Assay Details
Gene Symbol: SAMD4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062387.1 | XP_017917876.1 | 7 - 8 | 1828 | 63 | |
XM_018062388.1 | XP_017917877.1 | 8 - 9 | 1889 | 63 | ||
GenBank mRNA | JO546711.2 | - | 1018 | 63 | ||
KA336927.1 | - | 6 - 7 | 2006 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
