This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04756160_g1
See other ZYG11B GE Assays ›
Gene Symbol:
ZYG11B
Gene Name:
zyg-11 family member B, cell cycle regulator
Gene Aliases:
-
Chromosome Location:
Chr.3: 27209936 - 27281724 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04756160_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZYG11B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018044216.1 XP_017899705.1 5 - 6 1396 68
XM_018044224.1 XP_017899713.1 4 - 5 1292 68
GenBank mRNA JT687315.1 - 9 - 10 1788 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0