This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04753831_m1
See other ESRRA GE Assays ›
Gene Symbol:
ESRRA
Gene Name:
estrogen related receptor alpha
Gene Aliases:
-
Chromosome Location:
Chr.29: 43027000 - 43035204 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04753831_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESRRA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005699859.3 XP_005699916.1 4 - 5 1015 54
XM_018043293.1 XP_017898782.1 5 - 6 958 54
GenBank mRNA JO410161.2 - 5 - 6 948 54

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0