Assay Details
Gene Symbol: AK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045182.1 | XP_017900671.1 | 3 - 4 | 485 | 96 | |
XM_018045191.1 | XP_017900680.1 | 3 - 4 | 496 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
