This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04749676_m1
See other VAPA GE Assays ›
Gene Symbol:
VAPA
Gene Name:
VAMP associated protein A
Gene Aliases:
-
Chromosome Location:
Chr.24: 42254512 - 42292648 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04749676_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VAPA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005697139.3 XP_005697196.2 3 - 4 353 69
XM_018039547.1 XP_017895036.1 3 - 4 570 69
XM_018039548.1 XP_017895037.1 3 - 4 566 69
GenBank mRNA JO436626.2 - 3 - 4 755 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0