This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04749490_g1
See other PGAM2 GE Assays ›
Gene Symbol:
PGAM2
Gene Name:
phosphoglycerate mutase 2
Gene Aliases:
-
Chromosome Location:
Chr.22: 294628 - 296953 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04749490_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PGAM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005695487.3 XP_005695544.1 1 - 2 481 66
GenBank mRNA JO537975.2 - 2 - 3 608 66
KA347712.1 - 2 - 3 856 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

mutase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-6f7678f454-w8g9k:80/100.66.128.142:80.
git-commit: 7b0558793d0e70b0085ba95ba375cdfe261f8cbf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0