This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04749389_m1
See other HEXD GE Assays ›
Gene Symbol:
HEXD
Gene Name:
hexosaminidase D
Gene Aliases:
HEXDC
Chromosome Location:
Chr.19: 49689098 - 49706274 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04749389_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HEXD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005694373.3 XP_005694430.3 9 - 10 1827 91
XM_013972354.2 XP_013827808.2 9 - 10 1122 91
XM_018065400.1 XP_017920889.1 9 - 10 1122 91
GenBank mRNA JO399391.2 - 1 - 2 187 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0