This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04734885_g1
See other HDAC1 GE Assays ›
Gene Symbol:
HDAC1
Gene Name:
histone deacetylase 1
Gene Aliases:
-
Chromosome Location:
Chr.2: 14442095 - 14474909 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04734885_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018057759.1 XP_017913248.1 8 - 9 919 67
XM_018057766.1 XP_017913255.1 9 - 10 1052 67
GenBank mRNA JO499463.2 - 8 - 9 941 67
JO611119.2 - 6 - 7 1122 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0