This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04733085_m1
See other CKMT1A GE Assays ›
Gene Symbol:
CKMT1A
Gene Name:
creatine kinase, mitochondrial 1A
Gene Aliases:
CKMT1B
Chromosome Location:
Chr.21: 54833207 - 54839469 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04733085_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CKMT1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005695305.3 XP_005695362.1 9 - 10 1407 111
XM_018066415.1 XP_017921904.1 9 - 10 1713 111
XM_018066418.1 XP_017921907.1 9 - 10 1397 111
XM_018066419.1 XP_017921908.1 9 - 10 1414 111
GenBank mRNA KA309735.1 - 9 - 10 1500 111

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

amino acid kinase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0