Assay Details
Gene Symbol: CABP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013970799.2 | XP_013826253.1 | 5 - 6 | 1104 | 78 | |
XM_018061124.1 | XP_017916613.1 | 5 - 6 | 1199 | 78 | ||
XM_018061125.1 | XP_017916614.1 | 4 - 5 | 1001 | 78 | ||
GenBank mRNA | JO623764.2 | - | 2 - 3 | 263 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
