This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04727031_m1
See other CTAGE5 GE Assays ›
Gene Symbol:
CTAGE5
Gene Name:
CTAGE family member 5, ER export factor
Gene Aliases:
MIA2
Chromosome Location:
Chr.21: 48594423 - 48691238 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04727031_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTAGE5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018066279.1 XP_017921768.1 14 - 15 2701 75
XM_018066280.1 XP_017921769.1 13 - 14 2674 75
XM_018066281.1 XP_017921770.1 14 - 15 2702 75
XM_018066282.1 XP_017921771.1 13 - 14 2674 75
XM_018066284.1 XP_017921773.1 9 - 10 795 75
XM_018066285.1 XP_017921774.1 9 - 10 983 75
XM_018066286.1 XP_017921775.1 9 - 10 755 75
XM_018066287.1 XP_017921776.1 8 - 9 815 75
XM_018066288.1 XP_017921777.1 8 - 9 956 75
XM_018066289.1 XP_017921778.1 9 - 10 770 75
XM_018066290.1 XP_017921779.1 8 - 9 743 75
XM_018066291.1 XP_017921780.1 9 - 10 845 75
XM_018066292.1 XP_017921781.1 9 - 10 638 75
XM_018066293.1 XP_017921782.1 8 - 9 768 75
GenBank mRNA JO421670.2 - 16 - 17 1887 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0