Assay Details
Gene Symbol: SMPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058709.1 | XP_017914198.1 | 6 - 7 | 1627 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: SMPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058709.1 | XP_017914198.1 | 6 - 7 | 1627 | 114 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |