Assay Details
Gene Symbol: SMPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058709.1 | XP_017914198.1 | 4 - 5 | 1427 | 108 | |
GenBank mRNA | KA331772.1 | - | 2 - 3 | 169 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
