This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04721585_g1
See other RHOC GE Assays ›
Gene Symbol:
RHOC
Gene Name:
ras homolog family member C
Gene Aliases:
-
Chromosome Location:
Chr.3: 89966479 - 89972784 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04721585_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RHOC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018045922.1 XP_017901411.1 4 - 5 540 78
XM_018045923.1 XP_017901412.1 4 - 5 539 78
XM_018045924.1 XP_017901413.1 4 - 5 570 78
GenBank mRNA JO357775.2 - 1 - 2 542 78
JO576821.1 - 1 - 2 684 78
JO631976.2 - 1 - 2 636 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

small GTPase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0